Learn More
Description
Specifications
Specifications
| Antigen | HPV18 E7 |
| Applications | Western Blot, ELISA, KnockDown |
| Classification | Monoclonal |
| Clone | 718-15 |
| Conjugate | Alexa Fluor 647 |
| Dilution | Western Blot, ELISA, Sandwich ELISA, Knockdown Validated |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | E7 protein |
| Host Species | Mouse |
| Immunogen | Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with HPV oncoprotein E7, type 18. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
