Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV18 E7 Antibody (718-67cc), Novus Biologicals™
SDP

Catalog No. NB10073153 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10073153 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10073153 Supplier Novus Biologicals Supplier No. NB10073153
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Human Papilloma Virus Monoclonal specifically detects Human Papilloma Virus in Virus samples. It is validated for Sandwich ELISA, Western Blot.

Specifications

Antigen Human Papilloma Virus
Applications Sandwich ELISA, Western Blot
Classification Monoclonal
Clone 718-67cc
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Sandwich ELISA 1:100-1:2000
Formulation PBS (pH 7.4) with 0.09% Sodium Azide
Gene Alias E7, E7 protein, HPV16 E7, HpV16gp2, Human Papilloma Virus Type 18 E7 protein, Human papillomavirus E7 protein, Links Transforming protein human papillomavirus 16, Protein E7, Transforming protein, Transforming protein human papillomavirus 16
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with HPV oncoprotein E7, type 18.
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity HPV oncoprotein E7, type 18. Cross-reacts with type 16.
Target Species Virus
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.