Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPx1 Antibody (HIC0-3B3), Novus Biologicals™
SDP

Catalog No. NBP118951SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP118951SS 0.025 mL
NBP118951 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP118951SS Supplier Novus Biologicals Supplier No. NBP118951SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 9 publications

HPx1 Monoclonal specifically detects HPx1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen HPx1
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HIC0-3B3
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:100
Formulation Tissue culture supernatant with 0.05% Sodium Azide
Gene Alias human pancreatic exocrine cell
Host Species Mouse
Immunogen Human pancreatic enriched exocrine cells
Purification Method Tissue culture supernatant
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Pancreatic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Test Specificity Specific for exocrine cell types.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.