Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HR23A/Rad23A TUBE1 Antibody (SR1997), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32220350UL
Description
HR23A/Rad23A TUBE1 Monoclonal antibody specifically detects HR23A/Rad23A TUBE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
HR23A/Rad23A TUBE1 | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
5886 | |
Store at -20°C. Avoid freeze/thaw cycles. | |
IgG |
Western Blot, Flow Cytometry | |
SR1997 | |
Western Blot 1:1000-1:5000, Flow Cytometry 1:50 | |
hHR23A, HR23A | |
A synthesized peptide derived from human HR23A/Rad23A TUBE1 (Uniprot #: P54725) | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction