Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HR6A/UBE2A Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB291847 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291847 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291847 Supplier Novus Biologicals Supplier No. NB100555PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HR6A/UBE2A Polyclonal antibody specifically detects HR6A/UBE2A in Human samples. It is validated for Western Blot

Specifications

Antigen HR6A/UBE2A
Applications Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias EC 6.3.2.19, hHR6A, HR6A, RAD6ARAD6 homolog A, UBC2, Ubiquitin carrier protein A, ubiquitin-conjugating enzyme E2 A, ubiquitin-conjugating enzyme E2A (RAD6 homolog), Ubiquitin-protein ligase A
Host Species Rabbit
Immunogen A synthetic peptide which maps to a region between residues 100 and 150 of human ubiquitin-conjugating enzyme E2A using the numbering given in Swiss-Prot entry P49459 (GeneID 7319).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7319
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.