Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HR6B/UBE2B Antibody (OTI1E11), PerCP, Novus Biologicals™
SDP

Catalog No. NB288956 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288956 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288956 Supplier Novus Biologicals Supplier No. NBP271337PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HR6B/UBE2B Monoclonal antibody specifically detects HR6B/UBE2B in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen HR6B/UBE2B
Applications Western Blot
Classification Monoclonal
Clone OTI1E11
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias E2 protein, E2-17kDa, HHR6B, HR6B, RAD6 homolog B, RAD6BEC 6.3.2.19, UBC2, Ubiquitin carrier protein B, ubiquitin-conjugating enzyme E2 B, Ubiquitin-conjugating enzyme E2-17 kDa, ubiquitin-conjugating enzyme E2B (RAD6 homolog), Ubiquitin-protein ligase B
Host Species Mouse
Immunogen Full length human recombinant protein of human UBE2B (NP_003328) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 7320
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.