Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HR6B/UBE2B Antibody (PCRP-UBE2B-1C7), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB389472 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389472 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389472 Supplier Novus Biologicals Supplier No. NBP308912UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HR6B/UBE2B Monoclonal antibody specifically detects HR6B/UBE2B in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen HR6B/UBE2B
Applications Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-UBE2B-1C7
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias E2 protein, E2-17kDa, HHR6B, HR6B, RAD6 homolog B, RAD6BEC 6.3.2.19, UBC2, Ubiquitin carrier protein B, ubiquitin-conjugating enzyme E2 B, Ubiquitin-conjugating enzyme E2-17 kDa, ubiquitin-conjugating enzyme E2B (RAD6 homolog), Ubiquitin-protein ligase B
Host Species Mouse
Immunogen Recombinant full-length human HR6B/UBE2B protein (Uniprot: P63146)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 7320
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.