Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRD1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1002526 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002526 0.1 mL
NB1002526SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1002526 Supplier Novus Biologicals Supplier No. NB1002526
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 11 publications

HRD1 Polyclonal specifically detects HRD1 in Hamster, Human, Mouse samples. It is validated for Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen HRD1
Applications Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation reported in scientific literature (PMID 35350536), Immunohistochemistry-Paraffin 1:200 - 1:500. Use reported in scientific literature (PMID 24453213), Knockout Validated reported in scientific literature (PMID 26551274)
Formulation Whole antisera with No Preservative
Gene Accession No. Q86TM6
Gene Alias E3 ubiquitin-protein ligase synoviolin, EC 6.3.2, HRD1EC 6.3.2.-, KIAA1810Synovial apoptosis inhibitor 1, MGC40372, synovial apoptosis inhibitor 1, synoviolin
Gene Symbols SYVN1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of human HRD1 (within residues 300-400). [UniProt Q96PK3]
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 84447
Target Species Hamster, Mouse, Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.