Learn More
Description
Specifications
Specifications
| Antigen | HRD1 |
| Applications | Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation reported in scientific literature (PMID 35350536), Immunohistochemistry-Paraffin 1:200 - 1:500. Use reported in scientific literature (PMID 24453213), Knockout Validated reported in scientific literature (PMID 26551274) |
| Formulation | Whole antisera with No Preservative |
| Gene Accession No. | Q86TM6 |
| Gene Alias | E3 ubiquitin-protein ligase synoviolin, EC 6.3.2, HRD1EC 6.3.2.-, KIAA1810Synovial apoptosis inhibitor 1, MGC40372, synovial apoptosis inhibitor 1, synoviolin |
| Gene Symbols | SYVN1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
