Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRD1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB093560 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093560 100 μL
NB093561 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093560 Supplier Novus Biologicals Supplier No. NBP302998100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HRD1 Polyclonal specifically detects HRD1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen HRD1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias E3 ubiquitin-protein ligase synoviolin, EC 6.3.2, HRD1EC 6.3.2.-, KIAA1810Synovial apoptosis inhibitor 1, MGC40372, synovial apoptosis inhibitor 1, synoviolin
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 500 to the C-terminus of human HRD1 (NP_757385.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 84447
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.