Learn More
HRG beta-1 Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
$259.00 - $2255.00
Specifications
| Antigen | HRG beta-1 |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
500P28850UG
|
PeproTech
500P28850UG |
50 μg |
Each for $259.00
|
|
|||||
500P288100
|
PeproTech
500P288100UG |
100 μg |
Each for $331.00
|
|
|||||
500P2881MG
|
PeproTech
500P2881MG |
1 mg |
Each for $2,255.00
|
|
|||||
Description
AA Sequence of recombinant protein: SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Heregulinbeta-1. Anti-Human Heregulinbeta-1-specific antibody was purified by affinity chromatography employing an immobilized Human Heregulinbeta-1 matrix. Sandwich ELISA: To detect Human Heregulinbeta-1 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human Heregulinbeta- 1 (500-P288Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Heregulinbeta-1. Western Blot: To detect Human Heregulin-beta1 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human Heregulin-beta1 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Direct ligand for ERBB3 and ERBB4 tyrosine kinase receptors. Concomitantly recruits ERBB1 and ERBB2 coreceptors, resulting in ligand-stimulated tyrosine phosphorylation and activation of the ERBB receptors. The multiple isoforms perform diverse functions such as inducing growth and differentiation of epithelial, glial, neuronal, and skeletal muscle cells; inducing expression of acetylcholine receptor in synaptic vesicles during the formation of the neuromuscular junction; stimulating lobuloalveolar budding and milk production in the mammary gland and inducing differentiation of mammary tumor cells; stimulating Schwann cell proliferation; implication in the development of the myocardium such as trabeculation of the developing heart. Isoform 10 may play a role in motor and sensory neuron development.Specifications
| HRG beta-1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q02297 | |
| 3084 | |
| E.coli-derived Recombinant Human Heregulinbeta-1. | |
| Antigen affinity chromatography | |
| Nrg1 |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| 6030402G23Rik; acetylcholine receptor-inducing activity; ARIA; Breast cancer cell differentiation factor p45; D230005F13Rik; GGF; GGF2; GGFII; Glial growth factor; glial growth factor 2; heregulin; heregulin, alpha (45kD, ERBB2 p185-activator); Hgl; HRG; HRG1; HRGA; HRGalpha; MST131; MSTP131; NDF; Neu differentiation factor; neuregulin 1; neuregulin 1 type III beta 3; Neuregulin-1; Nrg1; NRG1-IT2; Pro-neuregulin-1, membrane-bound isoform; pro-NRG1; sensory and motor neuron derived factor; Sensory and motor neuron-derived factor; SMDF; type I neuregulin 1; type III neuregulin 1 | |
| Nrg1 | |
| Primary | |
| -20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.