Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hrk Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15944720UL
Description
Hrk Polyclonal specifically detects Hrk in Human samples. It is validated for Western Blot.Specifications
Hrk | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
O00198 | |
HRK | |
Synthetic peptides corresponding to HRK(harakiri, BCL2 interacting protein (contains only BH3 domain)) The peptide sequence was selected from the N terminal of HRK. Peptide sequence MCPCPLHRGRGPPAVCACSAGRLGLRSSAAQLTAARLKALGDELHQRTMW. | |
20 μL | |
Primary | |
Human | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
activator of apoptosis harakiri, activator of apoptosis Hrk, BCL2-interacting protein, BH3-interacting domain-containing protein 3, BID3, death protein 5, DP5, HARAKIRI, harakiri, BCL2 interacting protein (contains only BH3 domain), harakiri, BCL2-interacting protein (contains only BH3 domain), Neuronal death protein DP5 | |
Rabbit | |
Affinity Purified | |
RUO | |
8739 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction