Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hrk Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $499.50
Specifications
Antigen | Hrk |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15944720
![]() |
Novus Biologicals
NBP15944720UL |
20 μL |
Each for $206.00
|
|
|||||
NBP159447
![]() |
Novus Biologicals
NBP159447 |
100 μL |
Each for $499.50
|
|
|||||
Description
Hrk Polyclonal specifically detects Hrk in Human samples. It is validated for Western Blot.Specifications
Hrk | |
Polyclonal | |
Rabbit | |
O00198 | |
8739 | |
Synthetic peptides corresponding to HRK(harakiri, BCL2 interacting protein (contains only BH3 domain)) The peptide sequence was selected from the N terminal of HRK. Peptide sequence MCPCPLHRGRGPPAVCACSAGRLGLRSSAAQLTAARLKALGDELHQRTMW. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
activator of apoptosis harakiri, activator of apoptosis Hrk, BCL2-interacting protein, BH3-interacting domain-containing protein 3, BID3, death protein 5, DP5, HARAKIRI, harakiri, BCL2 interacting protein (contains only BH3 domain), harakiri, BCL2-interacting protein (contains only BH3 domain), Neuronal death protein DP5 | |
HRK | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title