Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRP-2 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB290108 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290108 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290108 Supplier Novus Biologicals Supplier No. NBP232074F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HRP-2 Polyclonal antibody specifically detects HRP-2 in Human samples. It is validated for Western Blot

Specifications

Antigen HRP-2
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias HDGF Like 2, HDGF2, HDGF-2, HDGFL2, HDGF-Related Protein 2, HDGFRP2, Hepatoma-derived growth factor 2, Hepatoma-Derived Growth Factor Related Protein 2, Hepatoma-Derived Growth Factor-Related Protein 2, HRP2
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 621 to 671 of human Hepatoma-Derived Growth Factor-Related Protein 2 using the numbering given in entry NP_001001520.1 (GeneID 84717).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 84717
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.