Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRP Antibody (2H11cc), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB359730 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359730 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359730 Supplier Novus Biologicals Supplier No. NB1108035AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HRP Monoclonal antibody specifically detects HRP in Nonspecies specific samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen HRP
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 2H11cc
Conjugate Alexa Fluor 350
Dilution ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias EC 1.11.1.7, Horseradish Peroxidase, HPRC 1, HPRC1, HRPA2, HRPC 2, HRPC2, HRPE5, HRPN, Neutral peroxidase, Peroxidase A2, Peroxidase C1A, Peroxidase C1B, Peroxidase C2, Peroxidase C3, Peroxidase E5, Peroxidase N, PRXC1A, PRXC1B, PRXC2, PRXC3
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified peroxidase from horseradish.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.