Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRP Antibody (HP-03), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB500420IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500420IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500420IR Supplier Novus Biologicals Supplier No. NB500420IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HRP Monoclonal specifically detects HRP in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HRP
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone HP-03
Conjugate DyLight 755
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 1.11.1.7, Horseradish Peroxidase, HPRC 1, HPRC1, HRPA2, HRPC 2, HRPC2, HRPE5, HRPN, Neutral peroxidase, Peroxidase A2, Peroxidase C1A, Peroxidase C1B, Peroxidase C2, Peroxidase C3, Peroxidase E5, Peroxidase N, PRXC1A, PRXC1B, PRXC2, PRXC3
Host Species Mouse
Immunogen Horseradish peroxidase (Grade I).
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The antibody HP-03 reacts with horseradish peroxidase. It is suitable for PAP detection system.
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.