Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HRPT2/CDC73 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4B4H2, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | HRPT2/CDC73 |
---|---|
Clone | 4B4H2 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6913
![]() ![]() |
Proteintech Group Inc
664901IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
U2 auxiliary factor (U2AF), comprised of a large and a small subunit, is a non-snRNP protein required for the binding of U2 snRNP to the pre-mRNA branch site. This gene encodes the U2AF large subunit which contains a sequence-specific RNA-binding region with 3 RNA recognition motifs and an Arg/Ser-rich domain necessary for splicing. The large subunit binds to the polypyrimidine tract of introns early during spliceosome assembly. Multiple transcript variants have been detected for this gene, but the full-length natures of only two have been determined to date.Specifications
HRPT2/CDC73 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C1orf28, CDC73, HPT JT, HRPT2, Hyperparathyroidism 2 protein, Parafibromin | |
CDC73 | |
IgG2a | |
Protein A | |
Antibody |
4B4H2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Pig, Rat | |
Q4V8C8, Q6P1J9, Q8JZM7 | |
100526068, 214498, 304832, 79577 | |
HRPT2; CDC73 Fusion Protein ag2959 | |
Primary | |
-20°C | |
CDC73 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title