Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRV3c Recognition Site Antibody, Novus Biologicals™
SDP

Catalog No. NBP229904 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229904 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229904 Supplier Novus Biologicals Supplier No. NBP229904
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HRV3c Recognition Site Polyclonal specifically detects HRV3c Recognition Site in Human, Canine, Baboon, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen HRV3c Recognition Site
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:2000, Immunoprecipitation 4 ug
Formulation PBS, 1.0 mg/ml BSA and 30% Glycerol with 0.05% Sodium Azide
Gene Alias HRV3c, Human Rhinovirus 3C, Rhinovirus 3C
Host Species Rabbit
Immunogen Synthetic peptide recognizing the cleavage site of HRV3c protease
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Canine, Baboon, Cynomolgus Monkey, Rhesus Macaque
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.