Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HS3ST3A1 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB182811 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB182811 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB182811 Supplier Novus Biologicals Supplier No. NBP306617R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HS3ST3A1 Polyclonal antibody specifically detects HS3ST3A1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen HS3ST3A1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias 3-OST-3A, 3OST3A1heparan sulfate glucosamine 3-O-sulfotransferase 3A1,30ST3A1heparin-glucosamine 3-O-sulfotransferase, EC 2.8.2, EC 2.8.2.30, h3-OST-3A, heparan sulfate (glucosamine) 3-O-sulfotransferase 3A1, Heparan sulfate 3-O-sulfotransferase 3A1, Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 3A1, HS3ST3A
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human HS3ST3A1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cytoskeleton Markers, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9955
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.