Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hsc70 Mouse anti-Human, Mouse, Rat, Clone: 1A4B3, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | Hsc70 |
|---|---|
| Clone | 1A4B3 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6868
![]()
|
Proteintech Group Inc
664421IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The product encoded by this gene belongs to the heat shock protein 70 family which contains both heat-inducible and constitutively expressed members. The latter are called heat-shock cognate proteins. This gene encodes a heat-shock cognate protein. This protein binds to nascent polypeptides to facilitate correct folding. It also functions as an ATPase in the disassembly of clathrin-coated vesicles during transport of membrane components through the cell. Two alternatively spliced variants have been characterized to date.Specifications
| Hsc70 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Heat shock 70 kDa protein 8, heat shock 70kDa protein 8, HSC54, Hsc70, HSC71, HSP71, HSP73, HSPA10, HSPA8, LAP1, NIP71 | |
| HSPA8 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1A4B3 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| P11142, P63017, P63018 | |
| 15481, 24468, 3312 | |
| Hsc70 Fusion Protein Ag17910 | |
| Primary | |
| -20°C | |
| HSPA8 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title