Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSCB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109833
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109833 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109833 Supplier Invitrogen™ Supplier No. PA5109833
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AASQAGSNYP RCWNCGGPWG PGREDRFFCP QCRALQAPDP TRDYFSLMDC NRSFRVDTAK LQHRYQQLQR LVHPDFFSQR SQTEKDFSEK HSTLVNDAYK TLLAPLSRGL YLLKLHGIEI PERTDYEMDR QFLIEIMEIN EKLAEAESEA AMKEIESIVK AKQKEFTDNV SSAFEQDDFE EAKEILTKMR YFSNIEEKIK LKKIPL.

May act as a co-chaperone in iron-sulfur cluster assembly in mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSCB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene HSCB
Gene Accession No. Q8IWL3
Gene Alias AI325508; AW049829; C-HSC20; dJ366L4.2; DnaJ (Hsp40) homolog, subfamily C, member 20; DnaJ homolog subfamily C member 20; DNAJC20; HSC20; HSCB; HscB iron-sulfur cluster co-chaperone homolog; HscB iron-sulfur cluster co-chaperone homolog (E. coli); HscB mitochondrial iron-sulfur cluster cochaperone; HscB mitochondrial iron-sulfur cluster co-chaperone; iron-sulfur cluster co-chaperone protein HscB; Iron-sulfur cluster co-chaperone protein HscB, cytoplasmic; Iron-sulfur cluster co-chaperone protein HscB, mitochondrial; JAC1; J-type co-chaperone HSC20; RGD1311005
Gene Symbols HSCB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-235 of human HSCB (NP_741999.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150274
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.