Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD17B2 Antibody (OTI4F10), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB270945APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270945APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270945APC Supplier Novus Biologicals Supplier No. NBP270945APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HSD17B2 Monoclonal specifically detects HSD17B2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen HSD17B2
Applications Flow Cytometry
Classification Monoclonal
Clone OTI4F10
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias 17-beta-hydroxysteroid dehydrogenase type 2, 20 alpha-hydroxysteroid dehydrogenase, E2DH, EC 1.1.1.62,17-beta-HSD 2, EC 1.1.1.63, EDH17B220-alpha-HSD, estradiol 17-beta-dehydrogenase 2, HSD17, hydroxysteroid (17-beta) dehydrogenase 2, Microsomal 17-beta-hydroxysteroid dehydrogenase, SDR9C2, short chain dehydrogenase/reductase family 9C, member 2, Testosterone 17-beta-dehydrogenase
Gene Symbols HSD17B2
Host Species Mouse
Immunogen Full length recombinant protein of human HSD17B2 (NP_002144) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3294.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.