Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD17B3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP216884 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216884 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216884 Supplier Novus Biologicals Supplier No. NBP216884
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HSD17B3 Polyclonal specifically detects HSD17B3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HSD17B3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.73mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS, 1% BSA, 20% Glycerol (pH7) with 0.025% Proclin 300
Gene Accession No. P37058
Gene Alias EC 1.1.1.64, EDH17B317-beta-HSD3, hydroxysteroid (17-beta) dehydrogenase 3,17-beta-hydroxysteroid dehydrogenase type 3, SDR12C2, short chain dehydrogenase/reductase family 12C, member 2, Testicular 17-beta-hydroxysteroid dehydrogenase, testosterone 17-beta-dehydrogenase 3,17-beta-HSD 3
Gene Symbols HSD17B3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human HSD17B3. The exact sequence is proprietary.
Molecular Weight of Antigen 35 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3293
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.