Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD17B8 Antibody (OTI2E12), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N271350A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271350A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271350A405 Supplier Novus Biologicals Supplier No. NBP271350AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HSD17B8 Monoclonal specifically detects HSD17B8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen HSD17B8
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2E12
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias D6S2245EdJ1033B10.9, EC 1.1.1.-, EC 1.1.1.62, EC 1.1.1.63,17-beta-HSD 8, estrogen 17-oxidoreductase, FABGLestradiol 17-beta-dehydrogenase 8, H2-KE6, HKE6FabG (beta-ketoacyl-[acyl-carrier-protein] reductase, E coli) like (E. coli), hydroxysteroid (17-beta) dehydrogenase 8,3-oxoacyl-[acyl-carrier-protein] reductase, ke-6, KE6, Ke-6,17-beta-hydroxysteroid dehydrogenase 8, Protein Ke6, Really interesting new gene 2 protein, RING2FABG, SDR30C1, short chain dehydrogenase/reductase family 30C, member 1, Testosterone 17-beta-dehydrogenase 8
Gene Symbols HSD17B8
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 57-261 of human HSD17B8 (NP_055049) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7923.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.