Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD3B1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB060076 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB060076 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB060076 Supplier Novus Biologicals Supplier No. NBP299334AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HSD3B1 Polyclonal specifically detects HSD3B1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen HSD3B1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1, 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I, 3BETAHSD, 3-beta-HSD I, 3-beta-hydroxy-Delta(5)-steroid dehydrogenase, 3BH, delta-5-3-ketosteroid isomerase, EC 1.1.1, HSD3B, HSDB3, HSDB3A, hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1, hydroxy-delta-5-steroid dehydrogenase, 3 beta-and steroid delta-isomerase 1, I, progesterone reductase, SDR11E1, short chain dehydrogenase/reductase family 11E, member 1,3-beta-hydroxy-5-ene steroid dehydrogenase, steroid Delta-isomerase, Trophoblast antigen FDO161G
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human HSD3B1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Prostate Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3283
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.