Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSD3B1, Mouse anti-Human, Clone: 4C1, Millipore Sigma™
Mouse Monoclonal Antibody
$192.34 - $498.79
Specifications
Antigen | HSD3B1 |
---|---|
Clone | 4C1 |
Dilution | Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected HSD3B1 in human adrenal gland tissue sections.ELISA Analysis : A representative lot detected HSD3B1 in ELISA applications (Gomez-Sanchez, C.E., et. al. (2017). Steroids. 127:56-61).Western Blotting Analysis: 1μg/mL from a representative lot detected HSD3B1 in human placenta tissue lysate in Western blotting applications.Immunohistochemistry (Paraffin) Analysis: A representative lot detected HSD3B1 in Immunohistochemistry applications (Gomez-Sanchez, C.E., et. al. (2017). Steroids. 127:56-61).Western Blotting Analysis: A representative lot detected HSD3B1 in Western Blotting applications (Gomez-Sanchez, C.E., et. al. (2017). Steroids. 127:56-61). |
Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABS206825
|
MilliporeSigma
MABS206825UG |
25 μg |
Each for $192.34
|
|
|||||
MABS2068100
|
MilliporeSigma
MABS2068100UG |
100 μg |
Each for $498.79
|
|
|||||
Description
Anti-HSD3B1, clone 4C1, Cat. No. MABS2068, is a mouse monoclonal antibody that detects 3-beta-HSD I (HSD3B1) and has been tested for use in ELISA, Immunohistochemistry (Paraffin), and Western Blotting.
3 beta-hydroxysteroid dehydrogenase/Delta 5--4-isomerase type 1 (UniProt: P14060; also known as 3 beta-hydroxysteroid dehydrogenase/Delta 5--4-isomerase type I, 3-beta-HSD I, Trophoblast antigen FDO161G) is encoded by the HSD3B1 (also known as 3BH, HSDB3A) gene (Gene ID: 3283) in human. HSD3B1 is a single -pass membrane protein localized in mitochondria and endoplasmic reticulum. Its transmembrane region is localized to amino acids 288-308. It is a bifunctional enzyme that catalyzes the oxidative conversion of Delta 5-ene-3-beta-hydroxy steroid and causes oxidative conversion of ketosteroids. It plays a vital role in the biosynthesis of all classes of hormonal steroids. Humans have two HSD3B isozymes. HSD3B1 is mainly expressed in the placenta, mammary and, prostate glands, and other peripheral tissues including the skin and adipose tissue and HSD3B2 is expressed primarily in the adrenal and gonads. They exhibit about 93% sequence homology. (Ref.: Gomez-Sanchez, CE et al. (2017). Steroids 127; 56-61).Specifications
HSD3B1 | |
Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected HSD3B1 in human adrenal gland tissue sections.ELISA Analysis : A representative lot detected HSD3B1 in ELISA applications (Gomez-Sanchez, C.E., et. al. (2017). Steroids. 127:56-61).Western Blotting Analysis: 1μg/mL from a representative lot detected HSD3B1 in human placenta tissue lysate in Western blotting applications.Immunohistochemistry (Paraffin) Analysis: A representative lot detected HSD3B1 in Immunohistochemistry applications (Gomez-Sanchez, C.E., et. al. (2017). Steroids. 127:56-61).Western Blotting Analysis: A representative lot detected HSD3B1 in Western Blotting applications (Gomez-Sanchez, C.E., et. al. (2017). Steroids. 127:56-61). | |
Monoclonal | |
Purified | |
RUO | |
Human | |
3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1;3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I;3-beta-HSD I;Trophoblast antigen FDO161G | |
HSD3B1;3BH;HSD3BA | |
IgG2b κ | |
Protein G purified | |
Clone 4C1 detects 3-beta-HSD I (HSD3B1) in human tissues. It targets an epitope with in 14 amino acids from the N-terminal region. |
4C1 | |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Signaling | |
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NP_000853.1 | |
A KLH-conjugated linear peptide corresponding to 14 amino acids from the N-terminal region of human 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1 (HSD3B1). | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title