Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSF1 Antibody (10H8), Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP1975040.2MG
Description
HSF1 Monoclonal specifically detects HSF1 in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HSF1 | |
Monoclonal | |
Unconjugated | |
PBS and 50% Glycerol with 0.09% Sodium Azide | |
HSF1 | |
Recombinant mouse HSF1. | |
0.2 mg | |
Primary | |
Human, Mouse, Rat, Primate, Rabbit | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
10H8 | |
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
Rat | |
Protein G purified | |
RUO | |
3297 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction