Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSF1 Antibody (10H8), Novus Biologicals™

Rat Monoclonal Antibody
$243.50 - $670.50
Specifications
Antigen | HSF1 |
---|---|
Clone | 10H8 |
Dilution | Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018794
![]() |
Novus Biologicals
NBP1975040.025MG |
0.025 mg |
Each for $243.50
|
|
|||||
NBP19750405
![]() |
Novus Biologicals
NBP1975040.05MG |
0.05 mg |
Each for $366.00
|
|
|||||
NBP19750402
![]() |
Novus Biologicals
NBP1975040.2MG |
0.2 mg |
Each for $670.50
|
|
|||||
Description
HSF1 Monoclonal specifically detects HSF1 in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HSF1 | |
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
PBS and 50% Glycerol with 0.09% Sodium Azide | |
3297 | |
Recombinant mouse HSF1. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
10H8 | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rat | |
Rabbit, Rat, Primate, Human, Mouse | |
heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
HSF1 | |
IgG1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title