Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSF1 Antibody (10H8), Novus Biologicals™
SDP

Rat Monoclonal Antibody

Supplier:  Novus Biologicals NBP1975040.05MG

 View more versions of this product

Catalog No. NBP19750405


Only null left
Add to Cart

Description

Description

HSF1 Monoclonal specifically detects HSF1 in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

HSF1
Monoclonal
Unconjugated
PBS and 50% Glycerol with 0.09% Sodium Azide
HSF1
Recombinant mouse HSF1.
0.05 mg
Primary
Human, Mouse, Rat, Primate, Rabbit
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
10H8
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1
Rat
Protein G purified
RUO
3297
Store at -20C. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only