Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSF1 Antibody (4B4), Biotin, Novus Biologicals™
SDP

Catalog No. NBP242206B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242206B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242206B Supplier Novus Biologicals Supplier No. NBP242206B
Add to Cart
Add to Cart

Rat Monoclonal Antibody

HSF1 Monoclonal specifically detects HSF1 in Human, Mouse, Rat, Bovine, Guinea Pig, Hamster, Monkey, Rabbit samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HSF1
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 4B4
Conjugate Biotin
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1
Gene Symbols HSF1
Host Species Rat
Immunogen Purified recombinant mouse HSF1 protein, epitope mapping to amino acids 425-439
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3297
Test Specificity Detects approx 85kDa (unstressed cell lysates) and approx 95kDa (heat shocked cell lysates).
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.