Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSF1 Antibody (5C7), Novus Biologicals™
SDP

Catalog No. NB298543 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298543 100 μL
NB298542 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298543 Supplier Novus Biologicals Supplier No. NBP326217100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HSF1 Monoclonal antibody specifically detects HSF1 in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen HSF1
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 5C7
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1
Host Species Rabbit
Immunogen A synthesized peptide derived from Human HSF1 [UniProt Q00613]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3297
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.