Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSF1 Antibody (5C7), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | HSF1 |
---|---|
Clone | 5C7 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
HSF1 Monoclonal antibody specifically detects HSF1 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
HSF1 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
A synthesized peptide derived from Human HSF1 [UniProt Q00613] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
5C7 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
3297 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title