Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP10/EPF Mouse anti-Bacteria, Alexa Fluor™ 594, Clone: M1.2, Novus Biologicals™
SDP

Catalog No. N1201327A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1201327A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1201327A94 Supplier Novus Biologicals Supplier No. NB12013527AF594

Mouse Monoclonal Antibody

HSP10/EPF Monoclonal specifically detects HSP10/EPF in Bacteria samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HSP10/EPF
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone M1.2
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 10 kDa chaperonin, 10 kDa heat shock protein, mitochondrial, Chaperonin 10, CPN10HSP10, Early-pregnancy factor, EPF, GROES, heat shock 10kD protein 1 (chaperonin 10), heat shock 10kDa protein 1 (chaperonin 10), Hsp10
Gene Symbols HSPE1
Host Species Mouse
Immunogen A 16 residue synthetic MAP peptide KKDDKGQQLPFEVQVG based on C. trachomatis cpn10 (residues 54-69).
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3336
Test Specificity This affinity purified detects a 15 kDa protein, corresponding to the apparent molecular mass of cpn10 on SDS-PAGE immunoblots, in C. trachomatis infected cells. This will not react with the human or mouse cpn10 homolog nor with E. coli GroES.
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.