Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSP20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583708
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583708 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583708 Supplier Invitrogen™ Supplier No. PA583708
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DQRFGEGLLE AELAALCPTT LAPYYLRAPS VALPVAQVPT DPGHFSVLLD VK Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 88%.

Heat Shock Protein 20 (HSP20), encoded by the HSPB6 gene, is a member of the small heat shock protein (sHSP) family, prominently involved in cellular stress response and maintenance of cytoskeletal integrity. HSP20 is ubiquitously expressed across various tissues, including cardiac and smooth muscle, where its roles are vital in modulating stress resilience and muscle function. It exerts protective effects against apoptosis and oxidative stress by interacting with cell signaling pathways that enhance cellular survival and protein stabilization. In cardiac muscle, HSP20 is known for its cardioprotective actions during ischemic injury, helping preserve myocardial function by inhibiting hypertrophic signaling pathways and promoting muscle relaxation. Studies highlight its involvement in vasodilation processes, as HSP20 interacts with myosin to regulate smooth muscle contraction and dilatation. Beyond its role in muscle physiology, HSP20 has been investigated for its potential therapeutic applications in cardiovascular diseases and metabolic disorders, where its expression and function could be pivotal in disease modulation and treatment strategies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP20
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HSPB6
Gene Accession No. O14558
Gene Alias AA387366; cardiac heat shock protein p20; cardiac p20; epididymis luminal protein 55; Gm479; Heat shock 20 kDa-like protein p20; heat shock 20-kDa protein; Heat shock protein; heat shock protein 20kDa; heat shock protein beta-6; heat shock protein family B (small) member 6; heat shock protein family B (small) member B6; heat shock protein, alpha-crystallin-related, B6; HEL55; HSP; HSP20; HSPB6; p20; PPP1R91; protein phosphatase 1, regulatory subunit 91
Gene Symbols HSPB6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HSPB6. Recombinant protein control fragment (Product #RP-98857).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126393
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.