Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP27 Antibody (017), Alexa Fluor™ 750
SDP

Catalog No. NB107665 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB107665 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB107665 Supplier Novus Biologicals Supplier No. NBP289442AF750

Rabbit Monoclonal Antibody

HSP27 Monoclonal specifically detects HSP27 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen HSP27
Applications Western Blot, ELISA
Classification Monoclonal
Clone 17
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human HSP27 (NP_001532.1; Met 1-Lys 205) .
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3315
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.