Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP27 Antibody
SDP

Catalog No. NB036606 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036606 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036606 Supplier Novus Biologicals Supplier No. NBP289015

Rabbit Polyclonal Antibody

HSP27 Polyclonal specifically detects HSP27 in Human, Bovine, Canine, Guinea Pig, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HSP27
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS containing 50% glycerol
Gene Alias 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a portion of human HSP27 phosphorylated at Ser15.
Purification Method Peptide affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3315
Target Species Human, Bovine, Canine, Guinea Pig, Monkey
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.