Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP27, p Ser78 Antibody (2079A) - Azide and BSA Free, R&D Systems™
SDP

Catalog No. MAB9355 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9355 100 μg
MAB9355SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB9355 Supplier Novus Biologicals Supplier No. MAB9355
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HSP27 (p Ser78) Monoclonal specifically detects HSP27 (p Ser78) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Simple Western, Western Blot.

Specifications

Antigen HSP27 (p Ser78)
Applications Flow Cytometry, Immunocytochemistry, Simple Western, Western Blot
Classification Monoclonal
Clone 2079A
Conjugate Unconjugated
Dilution Western Blot, Simple Western, Flow Cytometry 1-5 ug/ml, Immunocytochemistry
Formulation PBS with No Preservative
Gene Alias 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27
Gene Symbols HSPB1
Host Species Rabbit
Immunogen Phosphopeptide containing the human HSP27 S78 site Accession # P04792
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3315
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.