Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP27, p Ser78 Antibody (JBW502), Novus Biologicals™
SDP

Catalog No. NBP229821 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229821 0.1 mg
NB019478 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP229821 Supplier Novus Biologicals Supplier No. NBP229821
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HSP27 (p Ser78) Monoclonal specifically detects HSP27 (p Ser78) in Human samples. It is validated for Western Blot.

Specifications

Antigen HSP27 (p Ser78)
Applications Western Blot
Classification Monoclonal
Clone JBW502
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Gene Accession No. P04792
Gene Alias 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27
Gene Symbols HSPB1
Host Species Mouse
Immunogen peptide corresponding to amino acids 73-86 of human HSP27 with a C-terminus cysteine added for conjugation.
Molecular Weight of Antigen 27 kDa
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3315
Test Specificity Detects phosphorylated HSP27 (Ser78).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.