Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP27, p Ser78 Antibody (JBW502), Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $975.50
Specifications
Antigen | HSP27 (p Ser78) |
---|---|
Clone | JBW502 |
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
HSP27 Monoclonal specifically detects HSP27 in Human samples. It is validated for Western Blot.Specifications
HSP27 (p Ser78) | |
Western Blot 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27 | |
HSPB1 | |
IgG | |
Protein G purified | |
Detects phosphorylated HSP27 (Ser78). |
JBW502 | |
Western Blot | |
Unconjugated | |
Mouse | |
Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific | |
P04792 | |
3315 | |
peptide corresponding to amino acids 73-86 of human HSP27 with a C-terminus cysteine added for conjugation. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
27 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title