Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP40/DNAJB1 Antibody (2A7.H6), FITC, Novus Biologicals™
SDP

Catalog No. NBP122416F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP122416F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP122416F Supplier Novus Biologicals Supplier No. NBP122416F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HSP40/DNAJB1 Monoclonal specifically detects HSP40/DNAJB1 in Yeast samples. It is validated for ELISA.

Specifications

Antigen HSP40/DNAJB1
Applications ELISA
Classification Monoclonal
Clone 2A7.H6
Conjugate FITC
Dilution ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P25491
Gene Alias DnaJ (Hsp40) homolog, subfamily B, member 1, DnaJ (Hsp40) homolog, subfmaily B, member 1, dnaJ homolog subfamily B member 1, DnaJ protein homolog 1, DNAJ1, hDj-1, HDJ1, Heat shock 40 kDa protein 1, heat shock 40kD protein 1, Heat shock protein 40, HSP40, HSPF1Hdj1, Human DnaJ protein 1, radial spoke 16 homolog B, RSPH16B, Sis1
Gene Symbols DNAJB1
Host Species Mouse
Immunogen Full length protein HSP40 (YDJ1)
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline DNA Repair, Golgi Apparatus Markers, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 3337
Test Specificity Detects approx 40kDa. Yeast specific.
Target Species Yeast
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.