Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP40/DNAJB1 Antibody (OTI1F9), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP270969A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP270969A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP270969A70 Supplier Novus Biologicals Supplier No. NBP270969AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HSP40/DNAJB1 Monoclonal specifically detects HSP40/DNAJB1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HSP40/DNAJB1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1F9
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DnaJ (Hsp40) homolog, subfamily B, member 1, DnaJ (Hsp40) homolog, subfmaily B, member 1, dnaJ homolog subfamily B member 1, DnaJ protein homolog 1, DNAJ1, hDj-1, HDJ1, Heat shock 40 kDa protein 1, heat shock 40kD protein 1, Heat shock protein 40, HSP40, HSPF1Hdj1, Human DnaJ protein 1, radial spoke 16 homolog B, RSPH16B, Sis1
Gene Symbols DNAJB1
Host Species Mouse
Immunogen Full length recombinant protein of human HSP40/DNAJB1 (NP_006136) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA Repair, Golgi Apparatus Markers, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 3337.0
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.