Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp40/YDJ1 Antibody (1G10.H8), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP212879FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212879FR 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212879FR Supplier Novus Biologicals Supplier No. NBP212879FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hsp40/YDJ1 Monoclonal specifically detects Hsp40/YDJ1 in Yeast samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Hsp40/YDJ1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1G10.H8
Conjugate DyLight 680
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias MAS5, YDJ1 Ydj1p
Gene Symbols YDJ1
Host Species Mouse
Immunogen Full length protein yeast HSP40 (YDJ1)
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 855661
Test Specificity Detects approx 40kDa. Yeast specific product.
Target Species Yeast
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.