Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hsp47 Antibody (M16.10A1), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 14 publications
Supplier: Novus Biologicals NBP1974910.2MG
Description
Hsp47 Monoclonal specifically detects Hsp47 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Hamster, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.Specifications
| Hsp47 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Simple Western 1:100, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:10-1:500 | |
| (collagen binding protein 1), Arsenic-transactivated protein 3, AsTP3, CBP1, CBP2RA-A47, Cell proliferation-inducing gene 14 protein, Collagen-binding protein, colligen, Colligin, colligin-1, colligin-2, gp46, HSP47serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1), member 2, member 2, (collagen-binding protein 2), PPROM, proliferation-inducing gene 14, rheumatoid arthritis antigen A-47,47 kDa heat shock protein, Rheumatoid arthritis-related antigen RA-A47, serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), serpin H1, serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, SERPINH2colligin | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Recognizes Sheep HSP47. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| M16.10A1 | |
| Unconjugated | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| SERPINH1 | |
| Native rat HSP47. | |
| 0.2 mg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 871 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Guinea Pig, Hamster, Monkey, Rabbit, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction