Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp47 Antibody (M16.10A1), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 14 publications

$246.61 - $776.74

Specifications

Antigen Hsp47
Clone M16.10A1
Concentration 1.0 mg/mL
Dilution Western Blot 1:1000, Simple Western 1:100, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:10-1:500
Applications Immunohistochemistry, Immunofluorescence, Western Blot, Flow Cytometry, Immunocytochemistry
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018785
SDP
View Documents
Novus Biologicals
NBP1974910.025MG
0.025 mg
Each for $246.61
Only null left
Add to Cart
 
NBP19749105
SDP
View Documents
Novus Biologicals
NBP1974910.05MG
0.05 mg
Each for $444.80
Only null left
Add to Cart
 
NBP19749102
SDP
View Documents
Novus Biologicals
NBP1974910.2MG
0.2 mg
Each for $776.74
Only null left
Add to Cart
 
Description

Description

Hsp47 Monoclonal specifically detects Hsp47 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Hamster, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.
Specifications

Specifications

Hsp47
1.0 mg/mL
Immunohistochemistry, Immunofluorescence, Western Blot, Flow Cytometry, Immunocytochemistry
Unconjugated
Mouse
Core ESC Like Genes, Stem Cell Markers
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
871
Native rat HSP47.
Primary
Store at -20C. Avoid freeze-thaw cycles.
M16.10A1
Western Blot 1:1000, Simple Western 1:100, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:10-1:500
Monoclonal
Purified
RUO
Canine, Rat, Bovine, Sheep, Chicken, Human, Guinea Pig, Hamster, Mouse, Rabbit, Monkey, Pig
(collagen binding protein 1), Arsenic-transactivated protein 3, AsTP3, CBP1, CBP2RA-A47, Cell proliferation-inducing gene 14 protein, Collagen-binding protein, colligen, Colligin, colligin-1, colligin-2, gp46, HSP47serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1), member 2, member 2, (collagen-binding protein 2), PPROM, proliferation-inducing gene 14, rheumatoid arthritis antigen A-47,47 kDa heat shock protein, Rheumatoid arthritis-related antigen RA-A47, serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), serpin H1, serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, SERPINH2colligin
SERPINH1
IgG2b
Protein G purified
Recognizes Sheep HSP47.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.