Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp47 Rabbit anti-Human, Mouse, Clone: 3K3H1, Novus Biologicals™
SDP

Catalog No. NB165957 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165957 20 μg
NB165956 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB165957 Supplier Novus Biologicals Supplier No. NBP31543020UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Hsp47 Monoclonal antibody specifically detects Hsp47 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Hsp47
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3K3H1
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias (collagen binding protein 1), Arsenic-transactivated protein 3, AsTP3, CBP1, CBP2RA-A47, Cell proliferation-inducing gene 14 protein, Collagen-binding protein, colligen, Colligin, colligin-1, colligin-2, gp46, HSP47serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1), member 2, member 2, (collagen-binding protein 2), PPROM, proliferation-inducing gene 14, rheumatoid arthritis antigen A-47,47 kDa heat shock protein, Rheumatoid arthritis-related antigen RA-A47, serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), serpin H1, serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, SERPINH2colligin
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 319-418 of human Hsp47 (P50454).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 871
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.