Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP60 Antibody (2E1/53), Novus Biologicals™
SDP

Catalog No. NB1205479 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1205479 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1205479 Supplier Novus Biologicals Supplier No. NB1205479
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HSP60 Monoclonal specifically detects HSP60 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen HSP60
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2E1/53
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 - 10 ug/ml, ELISA 1 - 10 ug/ml, Immunohistochemistry 1 - 10 ug/ml, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunoprecipitation 2 ug, Immunohistochemistry-Paraffin 1 - 10 ug/ml
Gene Alias 60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant)
Gene Symbols HSPD1
Host Species Mouse
Immunogen Human placental HSP60.
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 3329
Test Specificity Detects Hsp 60 from human cells, tissues and recombinant protein preparations. This displays no other protein or species cross-reactivity.
Reconstitution Reconstitute with PBS.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.