Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP60 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP177397R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177397R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177397R Supplier Novus Biologicals Supplier No. NBP177397R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HSP60 Polyclonal specifically detects HSP60 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Knockdown Validated.

Specifications

Antigen HSP60
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot, KnockDown
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Knockdown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P10809
Gene Alias 60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant)
Gene Symbols HSPD1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human Hsp60 protein (within residues 70-150). [Swiss-Prot P10809]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cellular Markers, Membrane Trafficking and Chaperones, Mitochondrial Markers
Primary or Secondary Primary
Gene ID (Entrez) 3329
Test Specificity Human, mouse, rat, hamster and primate. Immunogen has 100% identity to bovine and chicken.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.