Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP60 Antibody (HSPD1/6496R), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB378214 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB378214 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB378214 Supplier Novus Biologicals Supplier No. NBP308938DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HSP60 Monoclonal antibody specifically detects HSP60 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Hamster, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen HSP60
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HSPD1/6496R
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias 60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant)
Host Species Rabbit
Immunogen Recombinant human full-length HSP60 protein (Uniprot: P10809)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cellular Markers, Loading Controls, Membrane Trafficking and Chaperones, Mitochondrial Markers
Primary or Secondary Primary
Gene ID (Entrez) 3329
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.