Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP70/DnaK Antibody (8E2/2), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP1974900.025MG
Description
HSP70/DnaK Monoclonal specifically detects HSP70/DnaK in Human, E. coli samples. It is validated for Western Blot.Specifications
HSP70/HSPA1A | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B | |
Mouse | |
70 kDa | |
0.025 mg | |
Cancer, Cellular Markers, Hypoxia, Membrane Trafficking and Chaperones | |
3303 | |
E. coli | |
Purified |
Western Blot | |
8E2/2 | |
Western Blot 1:10000 | |
P08107 | |
HSPA1A | |
E. coli DnaK. | |
Protein G purified | |
RUO | |
Primary | |
Recognizes Bacteria DnaK. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction