Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP70/DnaK Antibody (8E2/2), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00 - $743.50

Specifications

Antigen HSP70/HSPA1A
Clone 8E2/2
Dilution Western Blot 1:10000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018784
SDP
View Documents
Novus Biologicals
NBP1974900.025MG
0.025 mg
Each for $268.00
Only null left
Add to Cart
 
NBP19749005
SDP
View Documents
Novus Biologicals
NBP1974900.05MG
0.05 mg
Each for $426.00
Only null left
Add to Cart
 
NBP19749002
SDP
View Documents
Novus Biologicals
NBP1974900.2MG
0.2 mg
Each for $743.50
Only null left
Add to Cart
 
Description

Description

HSP70/DnaK Monoclonal specifically detects HSP70/DnaK in Human, E. coli samples. It is validated for Western Blot.
Specifications

Specifications

HSP70/HSPA1A
Western Blot 1:10000
Monoclonal
Purified
RUO
E. coli
P08107
3303
E. coli DnaK.
Primary
Store at -20C. Avoid freeze-thaw cycles.
70 kDa
8E2/2
Western Blot
Unconjugated
Mouse
Cancer, Cellular Markers, Hypoxia, Membrane Trafficking and Chaperones
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B
HSPA1A
IgG1
Protein G purified
Recognizes Bacteria DnaK.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.